Assay Details
Target Gene Details
Entrez Gene ID: | 114548 |
Gene Name: | NLR family pyrin domain containing 3 |
Gene Aliases: |
AGTAVPRL, AII, AVP, C1orf7, CIAS1, CLR1.1, FCAS, FCAS1, FCU, MWS, NALP3, PYPAF1 |
Location: |
Chr.1:247416156-247449108 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833137 | Chr.1:247274015 - 247444126 on Build GRCh38 | Gain |
|
| esv33129 | Chr.1:246911128 - 248165533 on Build GRCh38 | Gain+Loss |
|
| nsv1011097 | Chr.1:247402897 - 247503209 on Build GRCh38 | Gain |
|
| nsv509790 | Chr.1:247311000 - 247445876 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map