Assay Details
Target Gene Details
Entrez Gene ID: | 23624 |
Gene Name: | Cbl proto-oncogene C |
Gene Aliases: |
CBL-3, CBL-SL, RNF57 |
Location: |
Chr.19:44777855-44800646 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CBLC | NM_001130852.1 | NP_001124324.1 | ||
| NM_012116.3 | NP_036248.3 | |||
| XM_005258696.3 | XP_005258753.1 | |||
| XM_011526688.2 | XP_011524990.1 | |||
| XM_011526689.2 | XP_011524991.1 | |||
| XM_011526690.2 | XP_011524992.1 | |||
| AB028645.1 | BAA86298.1 | |||
| AF117646.1 | AAD34341.1 | |||
| AF117647.1 | AAD34342.1 | |||
| BC028915.1 | AAH28915.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833841 | Chr.19:44705217 - 44804786 on Build GRCh38 | Loss |
|
| nsv2506 | Chr.19:44750006 - 44794554 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map