Assay Details
Target Gene Details
Entrez Gene ID: | 7551 |
Gene Name: | zinc finger protein 3 |
Gene Aliases: |
A8-51, HF.12, KOX25, PP838, Zfp113 |
Location: |
Chr.7:100063847-100082548 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF3 | NM_001318137.1 | 4 | 1078 | NP_001305066.1 |
| NM_017715.3 | 6 | 1330 | NP_060185.2 | |
| AF193057.1 | 6 | 1293 | AAG22485.1 | |
| AF217988.1 | 6 | 1294 | AAG17231.1 | |
| AK000223.1 | 5 | 1273 | BAA91019.1 | |
| AK123799.1 | 1 | 1423 |
Target Gene Details
Entrez Gene ID: | 7589 |
Gene Name: | zinc finger and SCAN domain containing 21 |
Gene Aliases: |
NY-REN-21, ZNF38, Zipro1 |
Location: |
Chr.7:100049770-100065041 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZSCAN21 | NM_145914.2 | 4 | 1130 | NP_666019.1 |
| XM_005250568.4 | 5 | 2272 | XP_005250625.1 | |
| XM_006716113.3 | 4 | 2111 | XP_006716176.1 | |
| XM_017012584.1 | 5 | 2236 | XP_016868073.1 | |
| XM_017012585.1 | 5 | 1051 | XP_016868074.1 | |
| AF155100.1 | 3 | 805 | AAD42866.1 | |
| AK075324.1 | 6 | 1127 | ||
| AK292267.1 | 4 | 1150 | ||
| AK301871.1 | 6 | 1181 | ||
| AK313884.1 | 4 | 1148 | ||
| AL136865.1 | 3 | 1083 | CAB66799.1 | |
| BC033129.1 | 1 | 255 | AAH33129.1 | |
| BC047309.1 | 4 | 1123 | AAH47309.1 | |
| CR749301.1 | 5 | 1014 | CAH18156.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469687 | Chr.7:99974920 - 100157518 on Build GRCh38 | Loss |
|
| esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map