Assay Details
Target Gene Details
Entrez Gene ID: | 81577 |
Gene Name: | glucose-fructose oxidoreductase domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.16:67674533-67719370 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GFOD2 | NM_030819.3 | 3 | 1203 | NP_110446.3 |
| NR_027398.1 | 2 | 857 | ||
| XM_006721288.3 | 3 | 946 | XP_006721351.1 | |
| XM_017023738.1 | 3 | 1153 | XP_016879227.1 | |
| AK074382.1 | 4 | 1156 | BAB85066.1 | |
| AK091606.1 | 1 | 2080 | ||
| AK225997.1 | 3 | 1173 | ||
| AL832904.1 | 1 | 642 | CAH10637.1 | |
| AY358169.1 | 2 | 913 | AAQ88536.1 | |
| BC000757.2 | 2 | 810 | AAH00757.1 | |
| BC002943.1 | 2 | 830 | AAH02943.2 | |
| BC107585.1 | 3 | 1153 | AAI07586.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain |
|
| nsv952046 | Chr.16:67628798 - 67678697 on Build GRCh38 | Deletion |
|
| nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map