Assay Details
Target Gene Details
Entrez Gene ID: | 140687 |
Gene Name: | glucosaminyl (N-acetyl) transferase family member 7 |
Gene Aliases: |
C20orf105, dJ1153D9.2, gcnt |
Location: |
Chr.20:56491492-56525925 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCNT7 | NM_080615.1 | NP_542182.1 | ||
| AK131203.1 | BAD18395.1 |
Target Gene Details
Entrez Gene ID: | 51507 |
Gene Name: | replication termination factor 2 domain containing 1 |
Gene Aliases: |
C20orf43, CDAO5, HSPC164, SHUJUN-3 |
Location: |
Chr.20:56468585-56518886 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RTFDC1 | NM_001283035.1 | 10 | 1399 | NP_001269964.1 |
| NM_001283036.1 | 9 | 1306 | NP_001269965.1 | |
| NM_001283037.1 | 8 | 1254 | NP_001269966.1 | |
| NM_016407.4 | 9 | 1309 | NP_057491.2 | |
| XM_017027872.1 | 12 | 1502 | XP_016883361.1 | |
| AF117231.1 | 9 | 1257 | AAF17212.1 | |
| AF161513.1 | 9 | 1266 | AAF29128.1 | |
| AF161518.1 | 9 | 1235 | AAF29133.1 | |
| AF212244.1 | 9 | 1246 | AAK14929.1 | |
| AI352436.1 | 2 | 255 | ||
| AK000481.1 | 9 | 1239 | BAA91193.1 | |
| AK000586.1 | 8 | 1202 | ||
| AK026118.1 | 9 | 1237 | ||
| AK225135.1 | ||||
| AK302029.1 | 10 | 1342 | ||
| BC003359.1 | 9 | 1236 | AAH03359.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525425 | Chr.20:56403979 - 56897294 on Build GRCh38 | Gain |
|
| nsv1056574 | Chr.20:56495687 - 56761211 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map