Assay Details
Target Gene Details
Entrez Gene ID: | 7358 |
Gene Name: | UDP-glucose 6-dehydrogenase |
Gene Aliases: |
GDH, UDP-GlcDH, UDPGDH, UGD |
Location: |
Chr.4:39498755-39527598 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UGDH | NM_001184700.1 | NP_001171629.1 | ||
| NM_001184701.1 | NP_001171630.1 | |||
| NM_003359.3 | NP_003350.1 | |||
| XM_005262667.2 | XP_005262724.1 | |||
| AF049126.1 | AAC05135.1 | |||
| AF061016.1 | AAC36095.1 | |||
| AJ007702.1 | CAA07609.1 | |||
| AK097930.1 | ||||
| AK297737.1 | ||||
| BC022781.1 | AAH22781.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010189 | Chr.4:39423149 - 39523510 on Build GRCh38 | Gain |
|
| nsv1010784 | Chr.4:39437476 - 39571758 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map