Assay Details
Target Gene Details
Entrez Gene ID: | 90809 |
Gene Name: | transmembrane protein 55B |
Gene Aliases: |
C14orf9 |
Location: |
Chr.14:20457853-20461612 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM55B | NM_001100814.2 | 7 | 1540 | NP_001094284.1 |
| NM_144568.3 | 7 | 1519 | NP_653169.2 | |
| AL137727.1 | 2 | 3150 | CAB70896.1 | |
| BC002867.1 | 7 | 1301 | AAH02867.2 | |
| BC020947.1 | 7 | 1384 | AAH20947.1 | |
| BX161490.1 | 7 | 1385 | CAD61939.1 | |
| BX248025.1 | 7 | 1349 | CAD62347.1 | |
| CA430067.1 | 1 | 327 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv23438 | Chr.14:20332606 - 20789719 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map