Assay Details
Target Gene Details
Entrez Gene ID: | 10056 |
Gene Name: | phenylalanyl-tRNA synthetase beta subunit |
Gene Aliases: |
FARSLB, FRSB, HSPC173, PheHB, PheRS |
Location: |
Chr.2:222571443-222656355 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FARSB | NM_005687.4 | 17 | 2181 | NP_005678.3 |
| NR_130154.1 | 18 | 2377 | ||
| XM_006712169.2 | XP_006712232.1 | |||
| XM_011510466.2 | XP_011508768.1 | |||
| XM_017003110.1 | XP_016858599.1 | |||
| AF042346.1 | 17 | 1899 | AAD02220.1 | |
| AF161521.1 | AAF29136.1 | |||
| AK055289.1 | ||||
| AK291531.1 | ||||
| AK294158.1 | ||||
| AK316497.1 | ||||
| BC006502.2 | AAH06502.2 | |||
| BC017783.1 | AAH17783.1 | |||
| D84430.1 | 17 | 1912 | BAA95608.1 |
More Information
Set Membership: |
|

Genomic Map