Assay Details
Target Gene Details
Entrez Gene ID: | 401233 |
Gene Name: | HIV-1 Tat specific factor 1 pseudogene 2 |
Gene Aliases: |
- |
Location: |
Chr.6:3020156-3024771 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTATSF1P2 | NR_033884.1 | 1 | 1365 | |
| AK128391.1 | 1 | 490 | BAC87416.1 | |
| BX648511.1 | 1 | 485 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1029300 | Chr.6:2997310 - 3024828 on Build GRCh38 | Loss |
|
| nsv470795 | Chr.6:3019420 - 3058456 on Build GRCh38 | Gain |
|
| nsv5177 | Chr.6:2998306 - 3031324 on Build GRCh38 | Insertion |
|
| nsv830575 | Chr.6:2847376 - 3035682 on Build GRCh38 | Loss |
|

Genomic Map