Assay Details
Target Gene Details
Entrez Gene ID: | 83439 |
Gene Name: | transcription factor 7 like 1 |
Gene Aliases: |
TCF-3, TCF3 |
Location: |
Chr.2:85133460-85310388 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCF7L1 | NM_031283.2 | NP_112573.1 | ||
| XM_006712109.2 | XP_006712172.1 | |||
| AB031046.1 | BAB18185.1 | |||
| BC058894.1 | AAH58894.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834281 | Chr.2:85111413 - 85284344 on Build GRCh38 | Loss |
|
| esv3575266 | Chr.2:85065061 - 85379120 on Build GRCh38 | Gain |
|
| nsv582368 | Chr.2:85094623 - 85152929 on Build GRCh38 | Loss |
|
| nsv834279 | Chr.2:84980330 - 85178181 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map