Assay Details
Target Gene Details
Entrez Gene ID: | 1407 |
Gene Name: | cryptochrome circadian clock 1 |
Gene Aliases: |
PHLL1 |
Location: |
Chr.12:106991364-107093872 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRY1 | NM_004075.4 | NP_004066.1 | ||
| XM_017018832.1 | XP_016874321.1 | |||
| AK098615.1 | BAC05354.1 | |||
| AK125915.1 | ||||
| AK290552.1 | ||||
| BC030519.1 | AAH30519.1 | |||
| D83702.1 | BAA12068.1 | |||
| D84657.1 | BAA12710.1 |
More Information
Set Membership: |
|

Genomic Map