Assay Details
Target Gene Details
Entrez Gene ID: | 2444 |
Gene Name: | fyn related Src family tyrosine kinase |
Gene Aliases: |
GTK, PTK5, RAK |
Location: |
Chr.6:115941031-116100739 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FRK | NM_002031.2 | 8 | 2133 | NP_002022.1 |
| XM_005266880.4 | 9 | 2205 | XP_005266937.1 | |
| XM_005266881.2 | 9 | 2122 | XP_005266938.1 | |
| XM_005266882.4 | 10 | 2434 | XP_005266939.1 | |
| XM_011535653.2 | 10 | 2437 | XP_011533955.1 | |
| XM_011535654.2 | 9 | 2219 | XP_011533956.1 | |
| XM_011535655.2 | 9 | 2216 | XP_011533957.1 | |
| XM_011535656.2 | 8 | 1590 | XP_011533958.1 | |
| XM_017010645.1 | 9 | 2323 | XP_016866134.1 | |
| BC012916.1 | 8 | 1978 | AAH12916.1 | |
| U00803.1 | 8 | 2133 | AAA18284.1 | |
| U22322.1 | 8 | 2095 | AAC50116.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3610466 | Chr.6:115941729 - 115968686 on Build GRCh38 | Loss |
|
| nsv1029998 | Chr.6:115763322 - 116057888 on Build GRCh38 | Loss |
|
| esv3610465 | Chr.6:115941259 - 115955309 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map