Assay Details
Target Gene Details
Entrez Gene ID: | 50487 |
Gene Name: | phospholipase A2 group III |
Gene Aliases: |
GIII-SPLA2, SPLA2III, sPLA2-III |
Location: |
Chr.22:31134807-31140607 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G3 | NM_015715.4 | NP_056530.2 | ||
| XM_011530204.1 | XP_011528506.1 | |||
| XM_011530205.1 | XP_011528507.1 | |||
| AF220490.1 | AAF44746.1 | |||
| BC025316.1 | AAH25316.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829173 | Chr.22:31054057 - 31147970 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map