Assay Details
Target Gene Details
Entrez Gene ID: | 90780 |
Gene Name: | pygopus family PHD finger 2 |
Gene Aliases: |
1190004M21Rik |
Location: |
Chr.1:154957026-154961782 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PYGO2 | NM_138300.3 | 3 | 738 | NP_612157.1 |
| AF289598.1 | 3 | 490 | AAL55782.1 | |
| AF457208.1 | 3 | 532 | AAL91371.1 | |
| AK090545.1 | 1 | 56 | BAC03474.1 | |
| AK092389.1 | 3 | 687 | BAC03877.1 | |
| BC006132.1 | 3 | 704 | AAH06132.2 | |
| BC013725.1 | AAH13725.1 | |||
| BC032099.2 | 3 | 704 | AAH32099.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509502 | Chr.1:154904407 - 155142291 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map