Assay Details
Target Gene Details
Entrez Gene ID: | 4356 |
Gene Name: | membrane palmitoylated protein 3 |
Gene Aliases: |
DLG3 |
Location: |
Chr.17:43800799-43833192 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPP3 | NM_001932.4 | 20 | 2085 | NP_001923.2 |
| NR_003562.2 | 19 | 2064 | ||
| XM_006721915.2 | 21 | 2273 | XP_006721978.1 | |
| XM_011524828.2 | 20 | 2168 | XP_011523130.1 | |
| XM_017024656.1 | 20 | 2301 | XP_016880145.1 | |
| XM_017024657.1 | 19 | 1977 | XP_016880146.1 | |
| AB209232.1 | 3 | 4307 | BAD92469.1 | |
| AK311368.1 | 18 | 2161 | ||
| BC047017.1 | 19 | 2055 | AAH47017.1 | |
| BC056865.1 | 19 | 1999 | AAH56865.1 | |
| DB357996.1 | 1 | 324 | ||
| U37707.1 | 18 | 2246 | AAB36964.1 |
More Information
Set Membership: |
|

Genomic Map