Assay Details
Target Gene Details
Entrez Gene ID: | 10243 |
Gene Name: | gephyrin |
Gene Aliases: |
GEPH, GPH, GPHRYN, HKPX1, MOCODC |
Location: |
Chr.14:66507407-67735831 on Build GRCh38 |
Assay Gene Location: | Within Intron 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPHN | XM_017020925.1 | XP_016876414.1 |
Target Gene Details
Entrez Gene ID: | 51109 |
Gene Name: | retinol dehydrogenase 11 (all-trans/9-cis/11-cis) |
Gene Aliases: |
ARSDR1, CGI82, HCBP12, MDT1, PSDR1, RALR1, RDJCSS, SCALD, SDR7C1 |
Location: |
Chr.14:67676800-67695793 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RDH11 | NM_001252650.1 | 6 | 1557 | NP_001239579.1 |
| NM_016026.3 | 7 | 1767 | NP_057110.3 | |
| AB209223.1 | 4 | 3828 | BAD92460.1 | |
| AF151840.1 | 7 | 1717 | AAD34077.1 | |
| AF167438.1 | 7 | 1717 | AAF89632.1 | |
| AK057195.1 | 7 | 1603 | ||
| AK074749.1 | 7 | 1747 | ||
| BC000112.1 | 7 | 1684 | AAH00112.1 | |
| BC011727.2 | 7 | 1682 | AAH11727.1 | |
| BC026274.1 | 7 | 1704 | AAH26274.1 | |
| BC037302.1 | 7 | 1744 | AAH37302.1 | |
| BC051291.1 | 7 | 1677 | AAH51291.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521832 | Chr.14:67674292 - 67738159 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map