Assay Details
Target Gene Details
Entrez Gene ID: | 116931 |
Gene Name: | mediator complex subunit 12 like |
Gene Aliases: |
NOPAR, TNRC11L, TRALP, TRALPUSH |
Location: |
Chr.3:151085665-151436677 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED12L | NM_053002.5 | NP_443728.3 | ||
| XM_006713487.3 | XP_006713550.1 | |||
| XM_011512390.2 | XP_011510692.1 | |||
| XM_011512394.2 | XP_011510696.1 | |||
| XM_011512399.2 | XP_011510701.1 | |||
| XM_017005676.1 | XP_016861165.1 | |||
| XM_017005677.1 | XP_016861166.1 | |||
| XM_017005678.1 | XP_016861167.1 | |||
| XM_017005679.1 | XP_016861168.1 | |||
| XM_017005680.1 | XP_016861169.1 | |||
| AB046855.1 | BAB13461.1 | |||
| AF399708.1 | AAO38813.1 | |||
| AK128165.1 | ||||
| BC144559.1 | ||||
| BX108326.1 |
Target Gene Details
Entrez Gene ID: | 53829 |
Gene Name: | purinergic receptor P2Y13 |
Gene Aliases: |
FKSG77, GPCR1, GPR86, GPR94, P2Y13, SP174 |
Location: |
Chr.3:151326308-151329549 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2RY13 | NM_176894.2 | 2 | 512 | NP_795713.2 |
| XM_006713664.1 | 2 | 512 | XP_006713727.1 | |
| AF178982.1 | AAK18752.1 | |||
| AF295368.1 | 1 | 601 | AAK01864.1 | |
| AF345565.1 | 1 | 461 | AAK29068.1 | |
| AF406692.1 | 1 | 428 | AAL01038.1 | |
| AK313208.1 | 2 | 511 | ||
| BC029363.1 | 2 | 510 | ||
| BC041116.1 | 2 | 485 | AAH41116.2 | |
| BC066887.1 | 1 | 427 | AAH66887.1 | |
| DA676457.1 | ||||
| JF810883.1 | 2 | 491 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829757 | Chr.3:151327092 - 151508654 on Build GRCh38 | Loss |
|
| nsv954492 | Chr.3:151315313 - 151363512 on Build GRCh38 | Duplication |
|
| nsv592045 | Chr.3:151052827 - 151377305 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map