Assay Details
Target Gene Details
Entrez Gene ID: | 49861 |
Gene Name: | claudin 20 |
Gene Aliases: |
- |
Location: |
Chr.6:155264013-155276548 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLDN20 | NM_001001346.3 | 1 | 46 | NP_001001346.1 |
| BC020838.1 | 1 | 47 | AAH20838.1 |
Target Gene Details
Entrez Gene ID: | 51106 |
Gene Name: | transcription factor B1, mitochondrial |
Gene Aliases: |
CGI-75, CGI75, mtTFB, mtTFB1 |
Location: |
Chr.6:155229990-155314497 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TFB1M | NM_016020.3 | NP_057104.2 | ||
| XM_005267006.4 | XP_005267063.1 | |||
| XM_011535870.1 | XP_011534172.1 | |||
| XM_011535871.1 | XP_011534173.1 | |||
| XM_011535872.2 | XP_011534174.1 | |||
| XM_011535873.2 | XP_011534175.1 | |||
| XM_017010917.1 | XP_016866406.1 | |||
| AF151833.1 | AAD34070.1 | |||
| AK289484.1 | ||||
| BC005183.1 | ||||
| BC017788.1 | AAH17788.1 | |||
| BU849912.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1025003 | Chr.6:155077125 - 155755140 on Build GRCh38 | Gain |
|
| esv3611344 | Chr.6:155263573 - 155273392 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map