Assay Details
Target Gene Details
Entrez Gene ID: | 5697 |
Gene Name: | peptide YY |
Gene Aliases: |
PYY-I, PYY1 |
Location: |
Chr.17:43952733-44024260 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PYY | XM_011525035.1 | XP_011523337.1 |
Target Gene Details
Entrez Gene ID: | 84336 |
Gene Name: | transmembrane protein 101 |
Gene Aliases: |
- |
Location: |
Chr.17:44011188-44024260 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM101 | NM_001304813.1 | 5 | 1564 | NP_001291742.1 |
| NM_001304814.1 | 5 | 1661 | NP_001291743.1 | |
| NM_032376.3 | 4 | 1493 | NP_115752.1 | |
| XM_005257737.2 | 5 | 2841 | XP_005257794.1 | |
| XM_011525346.1 | 7 | 3078 | XP_011523648.1 | |
| XM_011525347.1 | 6 | 2955 | XP_011523649.1 | |
| XM_011525349.1 | 5 | 2861 | XP_011523651.1 | |
| XM_011525351.1 | 5 | 2308 | XP_011523653.1 | |
| XM_011525353.1 | 5 | 2720 | XP_011523655.1 | |
| AB097008.1 | 4 | 1429 | BAC77361.1 | |
| AI420766.1 | 1 | 109 | ||
| AK130810.1 | 4 | 1427 | ||
| AK172826.1 | 4 | 1445 | ||
| BC007438.2 | 4 | 1441 | AAH07438.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833454 | Chr.17:43940848 - 44158311 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map