Assay Details
Target Gene Details
Entrez Gene ID: | 10138 |
Gene Name: | YY1 associated factor 2 |
Gene Aliases: |
- |
Location: |
Chr.12:42157104-42238368 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YAF2 | NM_001190977.2 | 3 | 2939 | NP_001177906.1 |
| NM_001190979.2 | 5 | 3137 | NP_001177908.1 | |
| NM_001320080.1 | 5 | 3165 | NP_001307009.1 | |
| NM_005748.5 | 4 | 3065 | NP_005739.2 | |
| NR_135139.1 | 7 | 3307 | ||
| NR_135140.1 | 7 | 3339 |
More Information
Set Membership: |
|

Genomic Map