Assay Details
Target Gene Details
Entrez Gene ID: | 6768 |
Gene Name: | suppression of tumorigenicity 14 |
Gene Aliases: |
ARCI11, HAI, MT-SP1, MTSP1, PRSS14, SNC19, TADG15, TMPRSS14 |
Location: |
Chr.11:130159787-130210362 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST14 | NM_021978.3 | NP_068813.1 | ||
| AK314173.1 | ||||
| BM791311.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051470 | Chr.11:130125641 - 130195466 on Build GRCh38 | Gain |
|
| esv2759871 | Chr.11:130099327 - 130472822 on Build GRCh38 | Loss |
|
| nsv832295 | Chr.11:130099212 - 130260193 on Build GRCh38 | Gain |
|
| nsv517199 | Chr.11:130155050 - 130206439 on Build GRCh38 | Loss |
|
| nsv1143696 | Chr.11:130159705 - 130160205 on Build GRCh38 | Deletion |
|
| nsv556565 | Chr.11:129996758 - 130180927 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map