Assay Details
Target Gene Details
Entrez Gene ID: | 80063 |
Gene Name: | activating transcription factor 7 interacting protein 2 |
Gene Aliases: |
MCAF2 |
Location: |
Chr.16:10386055-10483640 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATF7IP2 | NM_001256160.1 | 4 | 1067 | NP_001243089.1 |
| NM_024997.3 | 3 | 1024 | NP_079273.2 | |
| NR_045815.1 | 2 | 956 | ||
| NR_045816.1 | 3 | 1024 | ||
| XM_006720953.3 | 2 | 851 | XP_006721016.1 | |
| XM_006720954.3 | 2 | 1990 | XP_006721017.1 | |
| XM_017023705.1 | 2 | 1992 | XP_016879194.1 | |
| XM_017023706.1 | 2 | 1992 | XP_016879195.1 | |
| XM_017023707.1 | 2 | 1992 | XP_016879196.1 | |
| XM_017023708.1 | XP_016879197.1 | |||
| XM_017023710.1 | 1 | 558 | XP_016879199.1 | |
| AK093414.1 | 4 | 1065 | BAC04157.1 | |
| AK296565.1 | ||||
| AK299040.1 | 3 | 1018 | ||
| AL832987.1 | 1 | 746 | CAH56323.1 | |
| AY560615.1 | 1 | 797 | AAT66299.1 | |
| BC033891.1 | 3 | 1023 | AAH33891.1 | |
| BC137079.1 | 2 | 923 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051406 | Chr.16:10365859 - 10570475 on Build GRCh38 | Loss |
|
| nsv519263 | Chr.16:10386989 - 10466528 on Build GRCh38 | Gain+Loss |
|
| nsv457405 | Chr.16:10388455 - 10466528 on Build GRCh38 | Loss |
|
| nsv1036128 | Chr.16:10404948 - 10470444 on Build GRCh38 | Loss |
|
| dgv2727n100 | Chr.16:9914166 - 10436036 on Build GRCh38 | Gain |
|
| nsv1040428 | Chr.16:10145408 - 11362136 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map