Assay Details
Target Gene Details
Entrez Gene ID: | 3145 |
Gene Name: | hydroxymethylbilane synthase |
Gene Aliases: |
PBG-D, PBGD, PORC, UPS |
Location: |
Chr.11:119084871-119093549 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HMBS | NM_000190.3 | NP_000181.2 | ||
| NM_001024382.1 | NP_001019553.1 | |||
| NM_001258208.1 | NP_001245137.1 | |||
| NM_001258209.1 | NP_001245138.1 | |||
| XM_005271531.1 | XP_005271588.1 | |||
| XM_005271532.1 | XP_005271589.1 | |||
| XM_005271533.3 | XP_005271590.1 | |||
| XM_011542796.1 | XP_011541098.1 | |||
| XM_017017629.1 | XP_016873118.1 | |||
| AK000628.1 | ||||
| AK131072.1 | ||||
| AK290275.1 | ||||
| AK309544.1 | ||||
| BC000520.2 | AAH00520.1 | |||
| BC008149.1 | AAH08149.1 | |||
| BC019323.1 | AAH19323.1 | |||
| BX647328.1 | ||||
| CB128006.1 | ||||
| X04217.1 | CAA27801.1 | |||
| X04808.1 | CAA28499.1 |
More Information
Set Membership: |
|

Genomic Map