Assay Details
Target Gene Details
Entrez Gene ID: | 6439 |
Gene Name: | surfactant protein B |
Gene Aliases: |
PSP-B, SFTB3, SFTP3, SMDP1, SP-B |
Location: |
Chr.2:85657317-85668741 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SFTPB | NM_000542.3 | 12 | 1563 | NP_000533.3 |
| NM_198843.2 | NP_942140.2 | |||
| XM_005264487.2 | 11 | 1522 | XP_005264544.1 | |
| XM_005264488.3 | 11 | 1431 | XP_005264545.2 | |
| XM_017004702.1 | XP_016860191.1 | |||
| XM_017004703.1 | XP_016860192.1 | |||
| AB439563.1 | ||||
| AB439564.1 | ||||
| AB439565.1 | 2 | 321 | ||
| AB439566.1 | 2 | 365 | ||
| AK303817.1 | ||||
| BC032785.1 | 11 | 1430 | AAH32785.1 | |
| CA439044.1 | ||||
| DQ317589.1 | ||||
| J02761.1 | 11 | 1440 | AAA60212.1 | |
| M16764.1 | 8 | 1142 | AAA88099.1 |
More Information
Set Membership: |
|

Genomic Map