Assay Details
Target Gene Details
Entrez Gene ID: | 5827 |
Gene Name: | peroxisomal membrane protein 2 |
Gene Aliases: |
PMP22 |
Location: |
Chr.12:132687606-132704991 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PXMP2 | NM_018663.2 | 5 | 769 | NP_061133.1 |
| AF250136.1 | 5 | 769 | AAF73963.1 | |
| AY044439.1 | 5 | 704 | AAK95858.1 | |
| BC009836.1 | 1 | 206 | AAH09836.1 | |
| BC073997.1 | 5 | 740 | AAH73997.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952512 | Chr.12:132192256 - 132711214 on Build GRCh38 | Deletion |
|
| esv3631309 | Chr.12:132688042 - 132708784 on Build GRCh38 | Gain |
|
| esv2422284 | Chr.12:132604248 - 132708921 on Build GRCh38 | Deletion |
|
| nsv1047500 | Chr.12:132666737 - 132763400 on Build GRCh38 | Gain |
|
| nsv1037074 | Chr.12:132613063 - 132801953 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map