Assay Details
Target Gene Details
Entrez Gene ID: | 5834 |
Gene Name: | phosphorylase, glycogen; brain |
Gene Aliases: |
GPBB |
Location: |
Chr.20:25248070-25298012 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PYGB | NM_002862.3 | 1 | 70 | NP_002853.2 |
| AB209080.1 | BAD92317.1 | |||
| AK299697.1 | 1 | 55 | ||
| AK309036.1 | 1 | 68 | ||
| BC017045.2 | 1 | 45 | AAH17045.1 | |
| BC030795.2 | AAH30795.1 | |||
| BQ433140.1 | 1 | 70 | ||
| J03544.1 | AAA59597.1 | |||
| U47025.1 | AAB60395.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv779e214 | Chr.20:25104610 - 25367702 on Build GRCh38 | Gain |
|
| nsv828708 | Chr.20:25243259 - 25250965 on Build GRCh38 | Gain |
|
| nsv522873 | Chr.20:23923336 - 26134874 on Build GRCh38 | Gain |
|
| nsv585746 | Chr.20:25090818 - 25489420 on Build GRCh38 | Gain |
|
| dgv4290n100 | Chr.20:25086650 - 25494993 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map