Assay Details
Target Gene Details
Entrez Gene ID: | 197021 |
Gene Name: | lactase like |
Gene Aliases: |
KLG, KLPH |
Location: |
Chr.15:66547468-66566025 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LCTL | NM_001278562.1 | NP_001265491.1 | ||
| NM_207338.3 | NP_997221.2 | |||
| NR_103739.1 | ||||
| XM_017021999.1 | XP_016877488.1 | |||
| AK090598.1 | ||||
| AK311441.1 | ||||
| AY358729.1 | AAQ89091.1 | |||
| EB361548.1 |
Target Gene Details
Entrez Gene ID: | 55055 |
Gene Name: | zwilch kinetochore protein |
Gene Aliases: |
KNTC1AP, hZwilch |
Location: |
Chr.15:66505083-66549485 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZWILCH | NM_001287821.1 | 19 | 2340 | NP_001274750.1 |
| NM_001287822.1 | 18 | 2244 | NP_001274751.1 | |
| NM_001287823.1 | 18 | 2193 | NP_001274752.1 | |
| NM_017975.4 | 19 | 2245 | NP_060445.3 | |
| AK023175.1 | 19 | 2235 | BAB14446.1 | |
| AK027468.1 | 10 | 1060 | BAB55133.1 | |
| BC036900.1 | 19 | 1991 | AAH36900.1 | |
| BX640701.1 | 19 | 2307 | CAE45821.1 |
More Information
Set Membership: |
|

Genomic Map