Assay Details
Target Gene Details
Entrez Gene ID: | 6898 |
Gene Name: | tyrosine aminotransferase |
Gene Aliases: |
- |
Location: |
Chr.16:71566851-71577095 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAT | NM_000353.2 | 2 | 215 | NP_000344.1 |
| AK313380.1 | 2 | 216 | ||
| BC020707.1 | 2 | 212 | AAH20707.1 | |
| BC130534.1 | 2 | 186 | ||
| X52520.1 | 2 | 212 | CAA36750.1 | |
| X55675.1 | 2 | 166 | CAA39210.1 |
More Information
Set Membership: |
|

Genomic Map