Assay Details
Target Gene Details
Entrez Gene ID: | 60313 |
Gene Name: | GC-rich promoter binding protein 1 like 1 |
Gene Aliases: |
SP192 |
Location: |
Chr.1:45627304-45686691 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPBP1L1 | NM_021639.4 | 6 | 1774 | NP_067652.1 |
| XM_017002019.1 | 6 | 1835 | XP_016857508.1 | |
| AF177334.1 | 4 | 592 | AAG17978.1 | |
| AK024972.1 | 4 | 581 | BAB15047.1 | |
| AK225197.1 | 4 | 581 | ||
| AL834194.1 | 4 | 1424 | CAD38884.1 | |
| BC015713.1 | 4 | 673 | AAH15713.1 | |
| BC022218.1 | 4 | 857 | AAH22218.1 | |
| BC052257.1 | 3 | 422 | AAH52257.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1014128 | Chr.1:45631998 - 45928868 on Build GRCh38 | Gain |
|
| esv3893512 | Chr.1:45476180 - 46068871 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map