Assay Details
Target Gene Details
Entrez Gene ID: | 3221 |
Gene Name: | homeobox C4 |
Gene Aliases: |
HOX3, HOX3E, cp19 |
Location: |
Chr.12:54016852-54056030 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOXC4 | NM_014620.5 | NP_055435.2 | ||
| BQ893403.1 | ||||
| X07495.1 | CAA30376.1 |
Target Gene Details
Entrez Gene ID: | 3222 |
Gene Name: | homeobox C5 |
Gene Aliases: |
CP11, HOX3, HOX3D |
Location: |
Chr.12:54016852-54035361 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOXC5 | NR_003084.2 |
Target Gene Details
Entrez Gene ID: | 3223 |
Gene Name: | homeobox C6 |
Gene Aliases: |
CP25, HHO.C8, HOX3, HOX3C |
Location: |
Chr.12:54016852-54030823 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOXC6 | NM_004503.3 | 2 | 818 | NP_004494.1 |
| NM_153693.4 | 3 | 1215 | NP_710160.1 | |
| AK314829.1 | ||||
| BC074844.2 | AAH74844.1 | |||
| BC074845.2 | AAH74845.1 | |||
| BF058345.1 | 1 | 279 | ||
| CR456954.1 | CAG33235.1 | |||
| M16938.1 | 3 | 942 | AAA36007.1 | |
| S82986.1 | 2 | 838 | AAB46892.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428282 | Chr.12:53953285 - 54140862 on Build GRCh38 | Gain |
|
| nsv826379 | Chr.12:53935085 - 54056069 on Build GRCh38 | Gain |
|
| nsv558998 | Chr.12:54025136 - 54034227 on Build GRCh38 | Loss |
|
| nsv832418 | Chr.12:54016556 - 54186660 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map