Assay Details
Target Gene Details
Entrez Gene ID: | 53831 |
Gene Name: | G protein-coupled receptor 84 |
Gene Aliases: |
EX33, GPCR4 |
Location: |
Chr.12:54362445-54365224 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR84 | NM_020370.2 | 2 | 994 | NP_065103.1 |
| XM_011538495.2 | 2 | 1681 | XP_011536797.1 | |
| AF237762.1 | 2 | 960 | AAK01857.1 | |
| AF282693.1 | 2 | 982 | AAF91467.1 | |
| AK290394.1 | 1 | 1528 | ||
| BC020614.1 | 2 | 992 | AAH20614.1 | |
| CD366881.1 | 1 | 532 | ||
| FJ348261.1 | 1 | 892 |
Target Gene Details
Entrez Gene ID: | 102724050 |
Gene Name: | uncharacterized LOC102724050 |
Gene Aliases: |
- |
Location: |
Chr.12:54353691-54467030 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724050 | NR_120486.1 | |||
| NR_120487.1 | ||||
| DA746386.1 | ||||
| DA746474.1 | ||||
| DA757660.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv559000 | Chr.12:54343917 - 54396183 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map