Assay Details
Target Gene Details
Entrez Gene ID: | 54942 |
Gene Name: | family with sequence similarity 206 member A |
Gene Aliases: |
C9orf6, CG-8, Simiate |
Location: |
Chr.9:108934393-108950745 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM206A | NM_017832.3 | NP_060302.1 | ||
| XM_011518816.2 | XP_011517118.1 | |||
| AF153417.1 | AAG43367.1 | |||
| AK000464.1 | BAA91183.1 | |||
| AK310297.1 | ||||
| BC015795.1 | AAH15795.1 |
More Information
Set Membership: |
|

Genomic Map