Assay Details
Target Gene Details
Entrez Gene ID: | 10694 |
Gene Name: | chaperonin containing TCP1 subunit 8 |
Gene Aliases: |
C21orf112, Cctq, D21S246, PRED71 |
Location: |
Chr.21:29056322-29073797 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCT8 | NM_001282907.1 | NP_001269836.1 | ||
| NM_001282908.1 | NP_001269837.1 | |||
| NM_001282909.1 | NP_001269838.1 | |||
| NM_006585.3 | NP_006576.2 | |||
| AK222490.1 | BAD96210.1 | |||
| AK293705.1 | ||||
| AK298801.1 | ||||
| AK308063.1 | ||||
| BC012584.1 | AAH12584.1 | |||
| BC072001.1 | AAH72001.1 | |||
| BC095470.1 | AAH95470.1 | |||
| D13627.1 |
More Information
Set Membership: |
|

Genomic Map