Assay Details
Target Gene Details
Entrez Gene ID: | 101928142 |
Gene Name: | AADACL2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:151751179-151928175 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AADACL2-AS1 | NR_110202.1 | |||
| NR_110203.1 | ||||
| DB454604.1 | ||||
| HY022483.1 | ||||
| HY205836.1 |
Target Gene Details
Entrez Gene ID: | 56670 |
Gene Name: | succinate receptor 1 |
Gene Aliases: |
GPR91 |
Location: |
Chr.3:151873643-151884619 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUCNR1 | NM_033050.5 | 3 | 1358 | NP_149039.2 |
| AF247785.1 | 4 | 1242 | AAL95690.1 | |
| AF348078.1 | 2 | 1295 | AAK29080.1 | |
| AK290420.1 | 3 | 1358 | ||
| BC030948.1 | 3 | 1352 | AAH30948.3 | |
| BI031507.1 | 1 | 320 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829758 | Chr.3:151849745 - 152005470 on Build GRCh38 | Gain |
|
| esv2759189 | Chr.3:151757492 - 151945387 on Build GRCh38 | Loss |
|
| nsv4067 | Chr.3:151879925 - 151910130 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map