Assay Details
Target Gene Details
Entrez Gene ID: | 7019 |
Gene Name: | transcription factor A, mitochondrial |
Gene Aliases: |
MTTF1, MTTFA, TCF6, TCF6L1, TCF6L2, TCF6L3 |
Location: |
Chr.10:58385143-58399230 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TFAM | NM_001270782.1 | NP_001257711.1 | ||
| NM_003201.2 | NP_003192.1 | |||
| NR_073073.1 | ||||
| AB451241.1 | ||||
| AB451366.1 | ||||
| AK312558.1 | ||||
| BC126366.1 | ||||
| BM455062.1 | ||||
| BT019658.1 | AAV38464.1 | |||
| BT019659.1 | AAV38465.1 | |||
| CR407653.1 | CAG28581.1 | |||
| EU279428.1 | ||||
| M62810.1 | AAA59849.1 |
More Information
Set Membership: |
|

Genomic Map