Assay Details
Target Gene Details
Entrez Gene ID: | 64092 |
Gene Name: | SAM domain, SH3 domain and nuclear localization signals 1 |
Gene Aliases: |
HACS1, NASH1, SASH2, SH3D6B, SLy2 |
Location: |
Chr.21:14485228-14583402 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAMSN1 | NM_001256370.1 | NP_001243299.1 | ||
| NM_001286523.1 | NP_001273452.1 | |||
| NM_022136.4 | NP_071419.3 | |||
| XM_011529684.2 | XP_011527986.1 | |||
| XM_011529685.1 | XP_011527987.1 | |||
| XM_011529686.1 | XP_011527988.1 | |||
| XM_017028427.1 | XP_016883916.1 | |||
| AF218085.2 | AAK07746.1 | |||
| AF222927.1 | AAG23355.1 | |||
| AF519621.1 | AAM75349.1 | |||
| AK125144.1 | ||||
| BC029112.1 | AAH29112.1 | |||
| HG380512.1 | ||||
| HG380514.1 |
More Information
Set Membership: |
|

Genomic Map