Assay Details
Target Gene Details
Entrez Gene ID: | 2131 |
Gene Name: | exostosin glycosyltransferase 1 |
Gene Aliases: |
EXT, LGCR, LGS, TRPS2, TTV |
Location: |
Chr.8:117797496-118111819 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXT1 | NM_000127.2 | 11 | 3202 | NP_000118.2 |
| AK130054.1 | 6 | 1979 | ||
| BC001174.1 | 11 | 2595 | AAH01174.1 | |
| S79639.1 | 11 | 3080 | AAB62283.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv820050 | Chr.8:117799541 - 117799766 on Build GRCh38 | Gain |
|
| nsv831438 | Chr.8:117667657 - 117811361 on Build GRCh38 | Loss |
|
| nsv831439 | Chr.8:117735113 - 117886879 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map