Assay Details
Target Gene Details
Entrez Gene ID: | 7026 |
Gene Name: | nuclear receptor subfamily 2 group F member 2 |
Gene Aliases: |
ARP1, CHTD4, COUPTFB, COUPTFII, NF-E3, NR2F1, SVP40, TFCOUP2 |
Location: |
Chr.15:96325928-96340263 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NR2F2 | NM_001145155.1 | 3 | 993 | NP_001138627.1 |
| NM_001145156.1 | 3 | 625 | NP_001138628.1 | |
| NM_001145157.1 | 3 | 1086 | NP_001138629.1 | |
| NM_021005.3 | 3 | 2234 | NP_066285.1 | |
| AB307711.1 | 3 | 1010 | ||
| AK298824.1 | 3 | 673 | ||
| AK301595.1 | 3 | 630 | ||
| AK316086.1 | 3 | 625 | ||
| BC014664.1 | 3 | 1160 | AAH14664.1 | |
| BC042897.1 | 4 | 1591 | AAH42897.1 | |
| BC106083.1 | 3 | 993 | AAI06084.1 | |
| FM208183.1 | 3 | 983 | ||
| HQ692848.1 | 3 | 1010 | ||
| HQ692849.1 | 3 | 611 | ||
| HQ692850.1 | 2 | 551 | ||
| M62760.1 | AAA21479.1 | |||
| M64497.1 | 3 | 1352 | AAA86429.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1141674 | Chr.15:95302056 - 96435607 on Build GRCh38 | Deletion |
|
| dgv448n67 | Chr.15:96307811 - 96377491 on Build GRCh38 | Gain |
|
| nsv827449 | Chr.15:96323551 - 96342897 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map