Assay Details
Target Gene Details
Entrez Gene ID: | 84695 |
Gene Name: | lysyl oxidase like 3 |
Gene Aliases: |
LOXL |
Location: |
Chr.2:74532258-74555709 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOXL3 | NM_001289164.1 | 11 | 1789 | NP_001276093.1 |
| NM_001289165.1 | 9 | 1506 | NP_001276094.1 | |
| NM_032603.3 | 13 | 2224 | NP_115992.1 | |
| XM_011533134.2 | 13 | 2184 | XP_011531436.1 | |
| XM_017005112.1 | 11 | 1828 | XP_016860601.1 | |
| AF282619.1 | 13 | 2197 | AAK51671.1 | |
| AF284815.1 | 13 | 2204 | AAK91134.1 | |
| AF311313.1 | 13 | 2195 | AAK63205.1 | |
| AK309474.1 | 11 | 1786 | ||
| AK309478.1 | 11 | 2405 | ||
| BC033130.1 | 6 | 730 | AAH33130.1 | |
| BC071865.1 | 11 | 1813 | AAH71865.1 | |
| DQ378059.1 | 9 | 1506 | ABD23013.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759061 | Chr.2:74252797 - 74535298 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map