Assay Details
Target Gene Details
Entrez Gene ID: | 1032 |
Gene Name: | cyclin dependent kinase inhibitor 2D |
Gene Aliases: |
INK4D, p19, p19-INK4D |
Location: |
Chr.19:10566462-10568979 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDKN2D | NM_001800.3 | 2 | 1095 | NP_001791.1 |
| NM_079421.2 | 3 | 841 | NP_524145.1 | |
| BC001822.2 | 3 | 825 | AAH01822.1 | |
| BI669755.1 | 3 | 846 | ||
| U40343.1 | 2 | 934 | AAB18139.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833744 | Chr.19:10458964 - 10635033 on Build GRCh38 | Loss |
|
| nsv833745 | Chr.19:10539132 - 10727920 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map