Assay Details
Target Gene Details
Entrez Gene ID: | 100500872 |
Gene Name: | microRNA 3911 |
Gene Aliases: |
mir-3911 |
Location: |
Chr.9:127690687-127690795 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR3911 | NR_037473.1 |
Target Gene Details
Entrez Gene ID: | 6812 |
Gene Name: | syntaxin binding protein 1 |
Gene Aliases: |
MUNC18-1, NSEC1, P67, RBSEC1, UNC18 |
Location: |
Chr.9:127612207-127692716 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STXBP1 | NM_001032221.3 | 19 | 1940 | NP_001027392.1 |
| NM_003165.3 | 20 | 2066 | NP_003156.1 | |
| AB209180.1 | 13 | 1204 | BAD92417.1 | |
| AF004562.1 | 19 | 1863 | AAC39688.1 | |
| AF004563.1 | 20 | 1989 | AAC39689.1 | |
| AK094794.1 | 1 | 682 | ||
| AK293984.1 | 18 | 1759 | ||
| AK295154.1 | 19 | 1924 | ||
| AK295802.1 | 15 | 1492 | ||
| BC015749.1 | 19 | 1856 | AAH15749.1 | |
| D63851.1 | 19 | 1812 | BAA19483.1 | |
| Y12723.1 | 19 | 1851 | CAA73255.1 |
More Information
Set Membership: |
|

Genomic Map