Assay Details
Target Gene Details
Entrez Gene ID: | 4953 |
Gene Name: | ornithine decarboxylase 1 |
Gene Aliases: |
ODC |
Location: |
Chr.2:10440371-10448554 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ODC1 | NM_001287188.1 | NP_001274117.1 | ||
| NM_001287189.1 | NP_001274118.1 | |||
| NM_001287190.1 | NP_001274119.1 | |||
| NM_002539.2 | NP_002530.1 | |||
| AK292352.1 | ||||
| AK301956.1 | ||||
| AK312766.1 | ||||
| BC025296.1 | AAH25296.1 | |||
| M16650.2 | AAA59966.2 | |||
| M20372.1 | AAA59968.1 | |||
| X55362.1 | CAA39047.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1007185 | Chr.2:10400260 - 10651617 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map