Assay Details
Target Gene Details
Entrez Gene ID: | 2648 |
Gene Name: | lysine acetyltransferase 2A |
Gene Aliases: |
GCN5, GCN5L2, PCAF-b, hGCN5 |
Location: |
Chr.17:42113111-42121393 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KAT2A | NM_021078.2 | NP_066564.2 | ||
| XM_006721817.3 | XP_006721880.1 | |||
| XM_006721818.3 | XP_006721881.1 | |||
| XM_017024448.1 | XP_016879937.1 | |||
| AF029777.1 | AAC39769.1 | |||
| BC032743.1 | AAH32743.1 | |||
| BC039907.1 | AAH39907.1 | |||
| BC105977.1 | AAI05978.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953898 | Chr.17:42113083 - 42131582 on Build GRCh38 | Deletion |
|
| nsv1056904 | Chr.17:42025823 - 42363204 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map