Assay Details
Target Gene Details
Entrez Gene ID: | 221264 |
Gene Name: | adenylate kinase 9 |
Gene Aliases: |
AK 9, AKD1, AKD2, C6orf199, C6orf224, dJ70A9.1 |
Location: |
Chr.6:109492855-109691253 on Build GRCh38 |
Assay Gene Location: | Within Exon 43 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AK9 | NM_001145128.2 | 36 | 4972 | NP_001138600.2 |
| XM_006715376.3 | 37 | 5837 | XP_006715439.2 | |
| XM_011535550.2 | 37 | 5852 | XP_011533852.1 | |
| XM_011535552.2 | 36 | 5028 | XP_011533854.1 | |
| XM_011535553.2 | 36 | 5635 | XP_011533855.1 | |
| XM_011535554.2 | 35 | 5531 | XP_011533856.1 | |
| XM_011535558.1 | 18 | 2873 | XP_011533860.1 | |
| XM_017010385.1 | 36 | 5020 | XP_016865874.1 | |
| XM_017010387.1 | 31 | 5483 | XP_016865876.1 | |
| XM_017010389.1 | 22 | 3735 | XP_016865878.1 | |
| AK098657.1 | 2 | 210 | BAC05368.1 | |
| BC101714.1 | 2 | 138 | AAI01715.1 | |
| BC111948.1 | 2 | 138 | AAI11949.1 | |
| BC143533.1 |
More Information
Set Membership: |
|

Genomic Map