Assay Details
Target Gene Details
Entrez Gene ID: | 6439 |
Gene Name: | surfactant protein B |
Gene Aliases: |
PSP-B, SFTB3, SFTP3, SMDP1, SP-B |
Location: |
Chr.2:85657317-85668741 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SFTPB | NM_000542.3 | 12 | 2707 | NP_000533.3 |
| NM_198843.2 | 12 | 1880 | NP_942140.2 | |
| XM_017004703.1 | XP_016860192.1 | |||
| AB439563.1 | 2 | 638 | ||
| AB439564.1 | ||||
| BG482852.1 | ||||
| BM989941.1 | 1 | 613 | ||
| CA435153.1 | 1 | 614 | ||
| CA437348.1 | ||||
| CA439044.1 | ||||
| DQ317589.1 |
More Information
Set Membership: |
|

Genomic Map