Assay Details
Target Gene Details
Entrez Gene ID: | 7113 |
Gene Name: | transmembrane protease, serine 2 |
Gene Aliases: |
PP9284, PRSS10 |
Location: |
Chr.21:41464551-41508158 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMPRSS2 | NM_001135099.1 | NP_001128571.1 | ||
| NM_005656.3 | NP_005647.3 | |||
| XM_005261043.2 | XP_005261100.1 | |||
| XM_011529731.2 | XP_011528033.1 | |||
| XM_011529732.2 | XP_011528034.1 | |||
| XM_011529733.1 | XP_011528035.1 | |||
| AF123453.1 | AAD37117.1 | |||
| AF270487.1 | AAK29280.1 | |||
| AF329454.1 | AAK53559.1 | |||
| AK222784.1 | BAD96504.1 | |||
| AK291813.1 | ||||
| AK296860.1 | ||||
| AK313338.1 | ||||
| BC051839.1 | AAH51839.1 | |||
| U75329.1 | AAC51784.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv459261 | Chr.21:41458763 - 41494180 on Build GRCh38 | Loss |
|
| nsv834102 | Chr.21:41286267 - 41494006 on Build GRCh38 | Loss |
|
| esv3647063 | Chr.21:41455441 - 41527402 on Build GRCh38 | Gain |
|
| esv3647064 | Chr.21:41456009 - 41569886 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map