Assay Details
Target Gene Details
Entrez Gene ID: | 55902 |
Gene Name: | acyl-CoA synthetase short-chain family member 2 |
Gene Aliases: |
ACAS2, ACECS, ACS, ACSA, dJ1161H23.1 |
Location: |
Chr.20:34874942-34927966 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACSS2 | NM_001242393.1 | NP_001229322.1 | ||
| XM_006723826.1 | XP_006723889.1 | |||
| XM_011528909.2 | XP_011527211.1 | |||
| XM_011528911.1 | XP_011527213.1 | |||
| XM_011528912.2 | XP_011527214.1 | |||
| DA626707.1 | ||||
| DA628275.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055447 | Chr.20:34737641 - 35020344 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map