Assay Details
Target Gene Details
Entrez Gene ID: | 8503 |
Gene Name: | phosphoinositide-3-kinase regulatory subunit 3 |
Gene Aliases: |
p55, p55-GAMMA, p55PIK |
Location: |
Chr.1:46040140-46176495 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3R3 | NM_001114172.1 | NP_001107644.1 | ||
| NM_001303427.1 | NP_001290356.1 | |||
| NM_001303428.1 | NP_001290357.1 | |||
| NM_001303429.1 | NP_001290358.1 | |||
| NM_003629.3 | NP_003620.3 | |||
| XM_017002614.1 | XP_016858103.1 | |||
| AF028785.1 | AAC39696.1 | |||
| AK302049.1 | ||||
| AK304713.1 | ||||
| AK313726.1 | ||||
| BC005381.1 | ||||
| BC021622.1 | AAH21622.1 | |||
| BX537388.1 | CAD97630.1 | |||
| CR749649.1 | CAH18443.1 | |||
| D88532.1 | BAA13636.1 | |||
| Y13809.1 | CAB52195.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3577805 | Chr.1:46003363 - 46075520 on Build GRCh38 | Gain |
|
| nsv1007750 | Chr.1:46005621 - 46086892 on Build GRCh38 | Gain |
|
| dgv31e212 | Chr.1:45920597 - 46063492 on Build GRCh38 | Gain |
|
| esv2761743 | Chr.1:45874186 - 46561071 on Build GRCh38 | Gain |
|
| esv3893512 | Chr.1:45476180 - 46068871 on Build GRCh38 | Gain |
|
| dgv19e214 | Chr.1:46030708 - 46154212 on Build GRCh38 | Gain |
|
| esv3585925 | Chr.1:45992755 - 46084727 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map