Assay Details
Target Gene Details
Entrez Gene ID: | 5025 |
Gene Name: | purinergic receptor P2X 4 |
Gene Aliases: |
P2X4, P2X4R |
Location: |
Chr.12:121209861-121234106 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2RX4 | NM_001256796.1 | NP_001243725.1 | ||
| NM_001261397.1 | NP_001248326.1 | |||
| NM_001261398.1 | NP_001248327.1 | |||
| NM_002560.2 | NP_002551.2 | |||
| NR_046372.1 | ||||
| NR_046373.1 | ||||
| XM_011538416.2 | XP_011536718.1 | |||
| AF000234.1 | AAB58405.1 | |||
| AK225604.1 | ||||
| AK297704.1 | ||||
| AK308868.1 | ||||
| BC033826.1 | AAH33826.1 | |||
| U83993.1 | AAD00553.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560455 | Chr.12:121087788 - 121363295 on Build GRCh38 | Gain |
|
| nsv1054878 | Chr.12:121082077 - 121328424 on Build GRCh38 | Gain |
|
| nsv560454 | Chr.12:121019270 - 121307939 on Build GRCh38 | Loss |
|
| nsv521928 | Chr.12:120603925 - 121356945 on Build GRCh38 | Gain |
|
| nsv560456 | Chr.12:121230451 - 121249371 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map