Assay Details
Target Gene Details
Entrez Gene ID: | 2805 |
Gene Name: | glutamic-oxaloacetic transaminase 1 |
Gene Aliases: |
AST1, ASTQTL1, GIG18, cAspAT, cCAT |
Location: |
Chr.10:99396870-99430773 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOT1 | NM_002079.2 | NP_002070.1 | ||
| AF052153.1 | AAC28622.1 | |||
| AK293483.1 | ||||
| AK301916.1 | ||||
| AK309174.1 | ||||
| AK312684.1 | ||||
| AY513279.1 | AAT08032.1 | |||
| BC000498.2 | AAH00498.1 | |||
| DA581933.1 | ||||
| M37400.1 | AAA35563.1 |
More Information
Set Membership: |
|

Genomic Map